Clever Culture Systems APAS Independence Instrument
clever-culture-banner-d-2395
clever-culture-product-0887
Automate your microbiology lab with the Clever Culture Systems APAS™ Independence instrument so you can free your valuable time for other critical testing tasks. It was the first automated culture plate reader with FDA clearance for in vitro diagnostic (IVD) use.
The Automated Plate Assessment System (APAS) uses imaging technology and artificial intelligence to scan agar plates, detect bacterial growth, and eliminate no growth and user-defined negative cultures from your workflow. It has been estimated that as much as 70% of urine cultures and close to 95% of Methicillin-resistant Staphylococcus aureus (MRSA) cultures are negative.
Read with Efficiency
The compact, standardized APAS instrument was designed to help you evaluate culture plates more quickly by sorting them into “significant,” “non-significant,” “negative,” and “for review” categories. The technology works rapidly, reduces the workload of trained microbiologists, and can swiftly determine which specimens require no further attention.
Operate with Convenience
APAS Independence is easy to use and compatible with your current streaking and incubation process. The unit measures 79 x 31.5 in. and sits on the floor to save bench space. The image analysis software — APAS Analysis Modules — are currently available for urine and MRSA cultures and can be used with cultures plated on Thermo Scientific™ SBA and MacConkey agars, Thermo Scientific™ Spectra™ chromogenic MRSA agar, BD SBA and MacConkey agars, and BD BBL CHROMagar™ MRSA II agar.
The APAS Independence is distributed in the U.S. and Europe by Thermo Fisher Scientific. Uses, applications, and availability of products in each country depends on local regulatory marketing authorization status. APAS™ is a trademark of Clever Culture Systems, which is the legal manufacturer of the APAS Independence.
Contact an Account Manager
Complete this form to learn more about the Clever Culture Systems APAS Independence instrument.
* = required field
Contact an Account Manager
Complete this form to learn more about the Clever Culture Systems APAS Independence instrument.
* = required field
thermo-fisher-scientific-logo-top-margin